EPM-2490
LOW VOLATILITY THERMALLY CONDUCTIVE SILICONE ADHESIVE
Description
  • Two-part, white, thermally conductive, electrically insulating, silicone system
  • Electrically insulative and non-corrosive
  • 15:1 Mix Ratio (Part A: Part B)
Applications
  • Flexible Thermal interface material (TIM) designed to provide heat transfer between electrical/electronic components and their heat sinks for a variety of component level packaging
  • For applications requiring a broad operating temperature range
  • Ideal where a form-in-place TIM is needed for use as a sealing, caulking, adhesive, or potting material in electronics applications requiring minimal volatility for prevention of contamination in sensitive devices
Product Details
Property Average Result
Thermal Conductivity 1.49 W/(mK)
Viscosity
Cure System
Tensile
Appearance
Durometer
Elongation
Flow Rate 0.3 in (7.62 mm)
Ionic Content Cl <3 ppm
Ionic Content K <3 ppm
Ionic Content Na <6 ppm
Lap Shear 160 psi (1.1 MPa)
Mix Ratio
Specific Gravity
Tack Free Time
Tear
Work Time 2 hours
Comment