Description
- Two-part, clear silicone gel
- Designed for enhanced performance in extreme low and high temperatures
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For applications requiring low volatility
- Useful for potting intricate assemblies due to a low viscosity
- As a low modulus embedding or potting compound for environmental protection of electronic assemblies and components
Product Details
Property | Average Result |
---|---|
Viscosity | |
Penetration | 3 mm |
Appearance | |
Cure | |
Cure System | |
Ionic Content Cl | <5 ppm |
Ionic Content K | <1 ppm |
Ionic Content Na | <4 ppm |
Mix Ratio | |
Volatile Content | |
Comment |