EPM-2420
LOW VOLATILITY GENERAL PURPOSE SILICONE ADHESIVE
Description
  • Two-part, clear silicone system
  • Incorporates an adhesion promoter
  • Low viscosity
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
  • Ideal for applications where clarity and low viscosity are important
  • For applications requiring low volatility
  • Use to adhere covers onto housings or for any application where grooves and other configurations require a material that self levels
Product Details
Property Average Result
Cure System
Viscosity
Durometer
Tensile
Elongation
Appearance
Cure
Ionic Content Cl <2 ppm
Ionic Content K <1 ppm
Ionic Content Na <8 ppm
Mix Ratio
Specific Gravity
Volatile Content
Comment