CV-1500
CONTROLLED VOLATILITY ELECTRICALLY CONDUCTIVE RTV SILICONE ADHESIVE/SEALANT
Description
- One-part, black, electrically conductive RTV silicone
- Non-corrosive and non-slumping
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use for RFI and EMI shielding in electrical and space applications requiring static dissipation
- Protects from extremes in temperature, humidity, radiation, thermal stress and mechanical stress
- Well suited for form-in-place conductive gaskets and attaching heating coils to solar collector plates
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Volume Resistivity | |
Cure System | |
Durometer | |
Tensile | |
Appearance | |
Cte | |
Cure | |
Elongation | |
Flow Rate | 0 in |
Lap Shear | 325 psi (2.24 MPa) |
Specific Gravity | |
Tack Free Time | |
Comment |