Description
- A solvent-free, thermosetting resin
- Forms a transparent, rigid material that retains its cured properties over a broad temperature range (up to 300°C/570°F)
- Has good dielectric properties at high temperatures and humidity
- Excellent resistance to radiation
- 100:1 Mix Ratio (CF-4721:Dicumyl Peroxide)
Applications
- For encapsulating electrical and electronic units
- For electronics and photonics applications
Product Details
Property | Average Result |
---|---|
Refractive Index | |
Viscosity | |
Mmol | 4.7 Vi/g |
Appearance | |
Cure | |
Durometer | |
Mix Ratio | |
Non Volatile Content | |
Specific Gravity | |
Volume Resistivity | |
Comment |