Description
- One-part acetoxy cured silicone elastomer dispersed in TertButyl Acetate
- Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance for moving, sliding, and rubbing parts
- A non-blocking topcoat for silicone surfaces that is easy to clean and prevents the accumulation of dirt, dust and foreign debris
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Suitable for dip casting and heat-curing of thin elastomeric films
- For use as an adhesive for bonding and sealing silicone together
- Low viscosity makes dispersions ideal for use as sprayable coatings
- Provides a RTV, low-friction coating on cured silicone substrates
- Ideal use for needle coating
NuSil Technology’s MED-6671 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property | Average Result |
---|---|
Cure System | Acetoxy |
Appearance | Translucent |
Cure | 24 hours / RTV |
Mix Ratio | 1 Part |
Comment | RTV, Low Coefficient of Friction Coating |
What is Medical Grade?
Discover how Medical-grade silicones are specifically designed, manufactured and purified to meet the strictest needs of the healthcare industry.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.