Description
- A unique granular solid
- Kosher and Halal certified
- Easily blends with other tableting ingredients
- Eliminates the cost and clean-up of applying liquid Silicone antiflatulents
- Contains all Compendial grade components suitable for oral use
- Can be combined with flavors and lubricants
- Produced in registered drug facilities according to criteria set by the U.S. Food and Drug Administration
Applications
- Cited in FDA regulations 21 CFR 332.10 and 21 CFR 332.15 as a safe and effective over-the-counter drug for use as an antiflatulent to alleviate the symptoms of gas associated with heartburn, sour stomach, acid indigestion and post operative gas pain
- Cited in FDA regulation 21 CFR 173.340 as being safe to use in food processing applications (refer to FDA regulation for allowable levels in specific food processing applications)
- For the manufacture of single- or double-layer tablets
- For forming uniform compositions in any concentration required
- For effective, stable antiflatulence, even in the presence of aluminum and magnesium hydroxide
Product Details
Property | Average Result |
---|---|
Simethicone Content | 30 % |
Defoaming Test | 3 seconds |
Appearance | |
Heavy Metals | 5 ppm |
Comment |