Description
- Designated by the FDA-OTC Antacid and Antiflatulent Review Panel as a safe and effective antiflatulent component of antacid and other gastrointestinal preparations
- Kosher and Halal certified
- Outstanding pharmaceutical versatility
- Excellent properties including low volatility and resistance to silica separation
- One of the fastest defoamers of any Simethicone made
- Produced in a registered drug facility according to criteria set by the U.S. Food and Drug Administration
- Demonstrates bubble-coalescing activity and lubricity
- Produced in a facility compliant with Good Manufacturing Practices (FDA regulations 21 CFR Part 210 & 211 and ICH Q7)
- Tested in accordance with and meets all the requirements of the United States Pharmacopeia (USP) for Simethicone and the European Pharmacopeia (EP) for Simeticone
Applications
- Cited in FDA regulations 21 CFR 332.10 and 21 CFR 332.15 as a safe and effective over-the-counter drug to alleviate the symptoms of gas associated with heartburn, sour stomach, acid indigestion and post operative gas pain
- Cited in FDA regulation 21 CFR 173.340 as being safe to use in food processing applications (refer to FDA regulation for allowable levels in specific food processing applications)
- Provides lubrication for uniform and bubble-free application of topical preparations such as antibiotic ointments and contraceptive gels
- Effective in acne and other dermatological ointments, creams and lotions due to its lubricity and anti-whitening properties
- As an antifoam in the manufacture and separation of drug products made by microbial fermentation
- For use in enzyme digestive aids and as a stabilizer in making belladonna extract and in cod liver oil extraction
- Useful in packaging aqueous solutions such as prepared douches where foaming can impede operations
- Listed in the International Cosmetic Ingredient Dictionary as a material useful for the formulation of cosmetic and toiletry products
- Valuable for packaging and filling operations for such products as liquid soaps, shampoos and hair conditioners
- For a soft, silky feel in many hand creams and lotions, as it provides better cost efficiency and performance than dimethicone
Product Details
Property | Average Result |
---|---|
Viscosity | |
Simethicone Content | 100 % |
Defoaming Test | 3 seconds |
Loss On Heating | 0.25 % |
Appearance | |
Heavy Metals | 5 ppm |
Comment |