Description
- Two-part, black electrically conductive RTV silicone
- 10:1 Mix Ratio (Part A:B)
Applications
- For applications requiring Ultra Low OutgassingTM and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- Use for RFI and EMI shielding in electronic and space applications
- Use to adhere covers onto housings or for any application where grooves and other configurations require a flowable to limited flow material
- For applications requiring a broader operating temperature range
Product Details
Property | Average Result |
---|---|
Volume Resistivity | 2.5 ohmcm |
Cure System | Platinum |
Durometer | 75 Type A |
Tensile | 475 psi (3.28 MPa) |
Appearance | Black |
Cure | 30 minutes / 150 °C |
Elongation | 90 % |
Lap Shear | 250 psi (1.72 MPa) |
Lap Shear | 250 psi (1.72 MPa) |
Mix Ratio | 10:1 |
Special Feature | |
Specific Gravity | 1.19 |
Work Time | 2 hours |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.