Description
- Two-part, pourable clear, silicone elastomer
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | 50 Type A |
Viscosity | 72,000 cP (72,000 mPa*s) |
Work Time | 50 hours |
Tensile | 730 psi (5.03 MPa) |
Appearance | Clear |
Cure | 30 minutes / 150 °C |
Cure System | Platinum |
Elongation | 390 % |
Mix Ratio | 1:1 |
Refractive Index | 1.41 |
Specific Gravity | 1.03 |
Tear | 80 ppi (14.11 kN/m) |
Comment | 1.41 R.I., Optically Clear, Pourable |
The Strongest Silicone Elastomer Ever Developed
Dr. Jim Lambert, NuSil® Director of Research and Development, Biomaterials, introduces our breakthrough material that has a tensile strength of up to 50% greater than existing silicone elastomers on the market.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.