Description
- A two-part, encapsulation gel
- Extended work time
- 1.46 refractive index for matching silica
- 1:1 Mix Ratio (Part A:B)
Applications
- For protection of sensitive photonics assemblies from mechanical shock, thermal shock, dust, and ambient atmosphere
- Extended work time allows voids in complex assemblies to fill in and permits time for any trapped air bubbles to float to the fluid surface and escape
- For applications requiring an operating temperature range of -40°C to 200°C
- (-40°F to 392°F) or for soldering operations up to 260°C (500°F) for 1 to 2 minutes
Product Details
Property | Average Result |
---|---|
Viscosity | 1,000 cP (1,000 mPa*s) |
Work Time | 8 hours |
Appearance | Clear |
Cure | 1 minutes / 65 °C |
Cure System | Platinum |
Durometer | 10 Type 00 |
Mix Ratio | 1:1 |
Operating Temperature | -40 °C / 200 °C |
Refractive Index | 1.46 |
Specific Gravity | 1.04 |
Comment | Firm Gel;Index Matches 1.46 RI such as fused silica. Will RTV within 24 hrs. 5 hr. minimum potlife based on 2X initial viscosity. |
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.