Description
- One-part silicone dispersion
- Provided as an RTV silicone dispersion in VM&P Naphtha
Applications
- For applications requiring extreme low temperature, low outgassing and minimal volatile condensables under extreme operating conditions
- As a protective overcoating for electronic components and atomic oxygen resistance in space applications
- Provides radiation and thermal stress resistance
- For applications requiring a broader operating temperature range
Meets or exceeds the ASTM E 595 low outgas specifications outlined in NASA SP-R-0022A and European Space Agency PSS-014-702, with a TML of ≤1% and CVCM of ≤0.1%
Product Details
Property | Average Result |
---|---|
Viscosity | 240 cP (240 mPa*s) |
Non Volatile Content | 60 % |
Appearance | Clear |
Tack Free Time | 50 minutes |
Cure | 7 days / RTV |
Cure System | Oxime |
Mix Ratio | 1 PART |
Refractive Index | 1.43 |
Specific Gravity | 1 |
Comment | 60% Solids, Atomic Oxygen Protective Overcoat |
What Is Space Grade?
Discover what it takes to make silicones worthy of the distinction "Space Grade" and the rigorous testing that these materials undergo.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.