My Account Menu
My Account Menu

PLY-7700 ETHOXY FUNCTIONAL DIMETHYLSILOXANE FLUID

 

 

  • Epoxy functional dimethylsiloxane low viscosity fluid

 

  • PLY-7700 is designed with ethoxy functional groups to be used for creating dimethylsiloxane containing polymeric structures or other applications where alkoxy functional groups are required.


 

PROPERTYAVERAGE RESULT
Volatile Content48 %
Viscosity6 cSt
AppearanceClear
Cure SystemNon-Curing
Mix Ratio1 PART
RheologyFlowable / Self Leveling
Special FeatureEthoxy functional
Specific Gravity0.95
CommentLow viscosity fluid