My Account Menu
My Account Menu
Implant Line

MED-6608-2 BLACK SILICONE COATING AND MARKING INK

Description

  • One-part, flowable silicone elastomer dispersion
  • Cures at room temperature upon exposure to atmospheric moisture
  • Non-corrosive cure system

Applications

  • Ideal for use as a marking ink for silicone parts and other components on which the coating must maintain long-term stability
  • For use in pad and transfer printing applications


Product Details

PROPERTYAVERAGE RESULT
Cure SystemCondensation
Viscosity1,150 cP (1,150 mPa*s)
Non Volatile Content70 %
Mix Ratio1 Part
AppearanceBlack
Cure7 days / RTV
Specific Gravity1.46
Tack Free Time80 minutes
CommentRTV Ink in Black

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application