Implant Line
MED-2310 RTV SILICONE FOAM
Description
- Two-part, room temperature curing, silicone foam
- Platinum cure
- No odor
- Yields a low density silicone foam when catalyzed
- 1:1 Mix Ratio (Part A: Part B)
Applications
- Can be used to make foam sheeting or ribbon
- Use in shock and vibration dampening situations requiring a lightweight, flexible foam
- Device applications requiring a low density component
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Viscosity | 40,000 cP (40,000 mPa*s) |
| Cure System | Platinum |
| Application Life | 2 minutes |
| Appearance | White |
| Mix Ratio | 1:1 |
| Comment | Low Density |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
