My Account Menu
My Account Menu
Premium Care Line

MED-9011 LOW CONSISTENCY SILICONE GREASE

Description

  • A low consistency dimethyl silicone grease
  • Non-curing
  • Available in a variety of packaging options for convenient dispensing and application

Applications

  • Effective high temperature lubricant for gaskets, o-rings, and other sealing mechanisms
  • Ideal for use in high vacuum/pressure systems on glass and ceramic joints


Product Details

PROPERTYAVERAGE RESULT
Volatile Content0.3 %
Viscosity95,000 cP (95,000 mPa*s)
AppearanceTranslucent
Non Volatile Content99.4 %
Specific Gravity1
Substrate
CommentLow Consistency Grease

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application