Premium Care Line
MED-6731 HIGH CONSISTENCY FLUOROSILICONE GREASE
Description
- A high consistency silicone grease containing dimethyl methyl-trifluoropropylsiloxane copolymer
- Non-curing
- Available in a variety of packaging options for convenient dispensing and application
Applications
- Effective high temperature lubricant for gaskets, o-rings, and other sealing mechanisms
- Ideal for use in high vacuum/pressure systems on glass and ceramic joints
- Useful in applications requiring solvent resistance
- For lubricating the surfaces of molded dimethyl silicone parts
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Volatile Content | 0.6 % |
| Viscosity | 2,000,000 cP (2,000,000 mPa*s) |
| Substrate | |
| Comment | High Consistency Methyl Fluoro Grease |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
