My Account Menu
My Account Menu
Implant Line

MED-366 SILICONE FLUID

Description

  • A transparent, colorless polydimethylsiloxane liquid
  • Available in standard viscosities of: 1,000 cP
  • Custom viscosities available upon request

Applications

  • For applications requiring a material that is highly water repellent and resists decomposition from heat and oxidation
  • Has good wetting properties and good lubricative characteristics in contact with plastics, rubber and skin


Product Details

PROPERTYAVERAGE RESULT
Volatile Content0.1 %
Viscosity1,000 cP (1,000 mPa*s)
AppearanceClear
Cure SystemNon-Curing
Mix Ratio1 Part
Refractive Index1.40
Specific Gravity0.97
CommentHighly purified Dimethyl Polymer

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application