Premium Care Line
MED10-6671 LOW COEFFICIENT OF FRICTION SILICONE COATING
Description
- One-part acetoxy cured silicone elastomer dispersed in TertButyl Acetate
- Cures to a thin silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance for moving, sliding, and rubbing parts
- A non-blocking topcoat for silicone surfaces that is easy to clean and prevents the accumulation of dirt, dust and foreign debris
- Cures at room temperature upon exposure to atmospheric moisture
Applications
- Low viscosity makes dispersions ideal for use as sprayable coatings
- For best results apply by spraying (dipping may also be utilized)
- Provides a RTV, low-friction coating on cured silicone substrates
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Cure System | Acetoxy |
| Appearance | Translucent |
| Cure | 24 hours / RTV |
| Mix Ratio | 1 Part |
| Comment | RTV, Low Coefficient of Friction Coating |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
