My Account Menu
My Account Menu
Implant Line

MED-6670 LOW COEFFICIENT OF FRICTION SILICONE COATING

Description

  • Two-part silicone elastomer dispersed in xylene
  • Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance
  • Cures via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)

Applications

  • Suitable for dip casting and heat-curing of thin elastomeric films
  • Low viscosity makes dispersions ideal for use as sprayable coatings
  • To provide a heat-cured, low-friction coating on cured silicone substrates
  • For tubing (ID/OD). Balloons, valves, stoppers, o-rings, and for moving or sliding parts


Product Details

PROPERTYAVERAGE RESULT
Cure SystemPlatinum
Non Volatile Content25 %
AppearanceTranslucent
Cure5 minutes / RTV
Mix Ratio1:1
Special Feature
Specific Gravity0.96
Work Time24 hours
CommentLow Coefficient of Friction Coating

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application