Premium Care Line
MED-4162 SILICONE DISPERSION
Description
- One-part, silicone dispersion containing high molecular weight hydroxyl-functional silicone polymer dispersed in xylene Non-curing
Applications
- Suitable for dip casting of thin elastomeric films
- Low viscosity makes dispersions ideal for use as sprayable coatings
- Ideal for use as a needle coating
- Provides excellent lubricity for cutting edges, needle cannula, and other applications requiring friction reduction
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Cure System | Non-Curing |
| Viscosity | 27,000 cP (27,000 mPa*s) |
| Non Volatile Content | 31 % |
| Appearance | Translucent |
| Mix Ratio | 1 PART |
| Solvent | Xylene |
| Special Feature | |
| Comment | Lubricious Coating |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
