My Account Menu
My Account Menu
Premium Care Line

MED-4159 SILICONE DISPERSION

Description

  • One-part, dispersion containing amino-functional silicone dispersed in a mixture of aliphatic or isopropanol solvents
  • Good adhesion to metal substrates
  • Cures at room temperature upon exposure to atmospheric moisture

Applications

  • Ideal for use as a silicone coating on metals and other substrates and also can be used as needle coatings
  • Provides excellent lubricity for cutting edges, needle cannula, and other applications requiring friction reduction


Product Details

PROPERTYAVERAGE RESULT
Cure SystemAmine
Viscosity200 cSt
Non Volatile Content53 %
AppearanceClear
Cure7 days / RTV
Mix Ratio1 PART
SolventStoddard Solvent & IPA
Special Feature
CommentLow Crosslink Density, Amino Functional Lubricious Silicone Coating

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application