Implant Line
MED50-4800-4 COLOR MASTERBATCH FOR LIQUID SILICONE ELASTOMERS
Description
- A translucent orange, single component masterbatch, with the consistency of a low viscosity paste
- Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
Applications
- For easy and more precise pigmentation of liquid silicone rubbers and low consistency elastomers suitable for injection molding
- Reduces production time and eliminates use of powders that may contaminate clean room environments
- Strict quality controls ensure color consistency
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Appearance | Translucent Orange |
| Mix Ratio | 1 PART |
| Comment |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
