PLY-7665 DIMETHYL DIPHENYL COPOLYMER
Description
- A 30 mole % diphenylsiloxane endblocked with reactive vinyl groups
Applications
- For curing with crosslinkers and catalysts to form adhesives, sealant, rubbers and gels due to their reactive nature
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Refractive Index | 1.52 |
| Viscosity | 575 cSt (619.14 mPa*s) |
| Mmol | 0.8 Vi/g |
| Appearance | Clear |
| Specific Gravity | 1.08 |
| Volatile Content | 0.1 % |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
