XL-7505 DIMETHYL SILICONE HYDRIDE TERMINATED CROSSLINKER
Description
- Dimethylsiloxane polymer, hydride terminated
Applications
- Commonly used for reacting with vinyl functional groups to form crosslinks in silicone systems to form adhesives, sealants, rubbers and gels.
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Refractive Index | 1.40 |
| Viscosity | 500 cP (500 mPa*s) |
| Mmol | 0.16 H/g |
| Appearance | Translucent |
| Volatile Content | 1 % |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
