XL-119 METHYL VINYL CYCLIC INHIBITOR
Description
- 2,4,6,8 –Tertramethyl-2,4,6,8-tetravinylcyclotetrasiloxane (95% minimum)
- CAS # 2254-06-5
Applications
- Competitive Inhibitor for adjusting cure rate of vinyl and hydride functional silicone systems that cure via hydrosilylation
- Compatible with a variety of silicone polymer systems for formulating silicone adhesives, elastomers and gels due to its reactivity and solubility with silicone
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Appearance | Clear |
| Cure System | Non-Curing |
| Mix Ratio | 1 PART |
| Mmol | 11.5 Vi/g |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
