CAT-102 PEROXIDE CATALYST
Description
- Peroxide catalyst masterbatch for use with high consistency silicone systems
- Di(2,4-dichlorobenzoyl)peroxide at 50% concentration in silicone oil
Applications
- To vulcanize silicone elastomer systems for calendaring, extrusion, or transfer and compression molding applications
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Appearance | White |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
