CAT-50 VINYL PLATINUM CATALYST
Description
- A platinum-divinyltetramethylsiloxane complex in a low molecular weight, functional vinyl polymer
Applications
- For RTV and accelerated heat-curing of vinyl-functional silicone systems
- For curing silicone polymer systems to form adhesives, sealant, rubbers and gels due to its reactive nature
- Designed for use with optically clear, high refractive index silicone elastomer systems
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Refractive Index | 1.40 |
| % Pt | 2.5 |
| Appearance | Light Amber |
| Specific Gravity | 0.98 |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
