My Account Menu
My Account Menu

CF1-141 HIGH TECHNOLOGY SILICONE PRIMER (RED)

Description

  • Specially formulated primer designed for use with platinum-cured systems where conventional silicone primers are insufficient
  • Colored red to provide easy visual determination of surface coverage
  • One-component primer supplied in Isopropyl Alcohol requires no mixing
  • Air-drying
  • Convenient container sizes produce less waste
  • Lower Volatile Organic Component (VOC) content

Applications

  • Improves the adhesion of addition-cured systems to various substrates including: metals, glass, ceramics, some plastics, and other silicone materials


Product Details

PROPERTYAVERAGE RESULT
Non Volatile Content6 %
Specific Gravity0.8
AppearanceRed
Cure SystemHydrolysis
CommentAddition Cure Systems, IPA Based, 6% Solids

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application