My Account Menu
My Account Menu

PSA-1180 PRESSURE SENSITIVE SILICONE ADHESIVE

Description

  • One-part, non-curing, pressure sensitive adhesive
  • Dispersed in ethyl acetate

Applications

  • For temporarily adhering various substrates including: metals, glass, silicone, organic rubbers and films


Product Details

PROPERTYAVERAGE RESULT
Cure SystemNon-Curing
Non Volatile Content70 %
Cure60 minutes / 85 °C
Viscosity3,300 cP (3,300 mPa*s)

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application