R-2615-3 RED POTTING AND ENCAPSULATING SILICONE ELASTOMER
Description
- Two-part, red, solvent free, low viscosity silicone
- Offers good physical and electrical stability across a broad range of temperatures
- Convenient 1:1 mix ratio
- 1:1 Mix Ratio (Part A:Part B)
Applications
- To provide protection of electronic components and assemblies against shock, vibration, moisture, ozone, dust, chemicals and other environmental hazards by potting or encapsulating
- For applications requiring an operating temperature range of –65 to 240°C (-85 to 465°F)
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Viscosity | 4,250 cP |
| Durometer | 50 Type A |
| Tensile | 1,000 psi (6.9 MPa) |
| Elongation | 100 % |
| Appearance | Red |
| Cure | 30 minutes / 150 °C |
| Dielectric Strength | 830 volts/mil (32.37 kV/mm) |
| Specific Gravity | 1.06 |
| Work Time | 2 hours |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
