CF19-2615 OPTICALLY CLEAR POTTING AND ENCAPSULATING SILICONE ELASTOMER
Description
- Two-part, optically clear, solvent free, low viscosity silicone
- Cures at room temperature or rapidly with heat
- Offers good physical and electrical stability across a broad range of temperatures
- Convenient 1:1 Mix ratio
Applications
- To provide protection of electronic components and assemblies against shock, vibration, moisture, ozone, dust, chemicals and other environmental hazards by potting or encapsulating
- For photonics applications such as HBLEDs, photo detectors, lasers, discrete optics and flat panel displays that require a 1.41 refractive index
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Cure System | Platinum |
| Viscosity | 1,300 cP (1,300 mPa*s) |
| Durometer | 30 Type A |
| Tensile | 120 psi (0.83 MPa) |
| Elongation | 100 % |
| Appearance | Clear |
| Cure | 30 minutes / 150 °C |
| Dielectric Strength | 500 volts/mil (19.5 kV/mm) |
| Mix Ratio | 1:1 |
| Refractive Index | 1.41 |
| Volume Resistivity | 1,000,000,000,000,000 ohmcm |
| Work Time | 4 hours |
| Comment | Versatile, clear coating, cure can be heat accelerated. |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
