My Account Menu
My Account Menu

R-2100-2 BLACK SILICONE COATING / MARKING INK

Description

  • Two-part, addition-cure black silicone ink dispersed in xylene
  • Cures rapidly with heat

Applications

  • For use as a marking ink for silicone rubber parts and other components
  • Use in applications requiring a rapid cure
  • Various application methods including pad printing and screen printing


Product Details

PROPERTYAVERAGE RESULT
Cure SystemPlatinum
Viscosity900 cP (900 mPa*s)
Non Volatile Content60 %
Mix Ratio1:1
AppearanceBlack
Cure5 minutes / 150 °C
Tack Free Time24 hours
CommentAvailable in white (R-2100-1), black (R-2100-2), red (R-2100-3), and green (R-2100-6). Dispersed in xylene.

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application