CV-9042 THERMALLY CONDUCTIVE, CONTROLLED VOLATILITY SILICONE GREASE
Description
- Thermally conductive, white silicone grease
- Excellent resistance to moisture, ozone and oxidation over a wide temperature range
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions to avoid condensation in sensitive devices
- For thermal conductivity, high and low temperature stability, and low bleed
- Apply to the base and mounting studs of transistors and other electrical components as a heat-sink seal
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Specific Gravity | 1.61 |
| Appearance | White |
| Comment | Thermally Conductive |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
