LS-1246 THIXOTROPIC OPTICAL SILICONE GREASE
Description
- A thixotropic, one-part, ultra-clear optical grease comprised of an optical grade fluid immobilized in a nanoparticle powder
- Flows under pressure but non-slumping at rest
- Low modulus
- Non-curing
- 1.46 refractive index for matching silica
- Supplied in syringes for ease of dispensing
- Container options are available for use with automated dispensing equipment
Applications
- Coupling of silica telecom fiber splices, polymer waveguide/backplane connections, Vertical-Cavity Surface-Emitting Laser (VCSEL) coupling to fiber and a variety of other optics applications
- Optically clear matrix
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Specific Gravity | 1.1 |
| Appearance | Clear |
| Refractive Index | 1.46 |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
