My Account Menu
My Account Menu

LS1-3443 OPTICALLY CLEAR SILICONE GEL

Description

  • A two-part optically clear gel
  • A 1.43 refractive index at 589 nm
  • 1:1 Mix Ratio (Part A:B)

Applications

  • For applications requiring index matching at 1.43
  • For protection of sensitive photonics assemblies from mechanical shock, thermal shock, dust, and ambient atmosphere


Product Details

PROPERTYAVERAGE RESULT
Viscosity2,500 cP (2,500 mPa*s)
Penetration6 mm
AppearanceClear
Specific Gravity1

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application