My Account Menu
My Account Menu

CV-2391 CONTROLLED VOLATILITY RTV SILICONE FOAM

Description

  • Yields a low to medium density silicone rubber foam when rapidly catalyzed
  • Low viscosity
  • 1:10 Mix Ratio (Part A: Part B)

Applications

  • In shock and vibration dampening situations requiring a lightweight, flexible foam
  • For intricate back-filling due to a low viscosity
  • For applications requiring low outgassing and minimal volatile condensable materials.


Product Details

PROPERTYAVERAGE RESULT
Viscosity3,000 cP (3,000 mPa*s)
Cure SystemPlatinum
Foam Density20 lbs/ft3 (0.32 g/cm3)
Application Life6 minutes
AppearanceWhite
Cure60 minutes / RTV
Mix Ratio1:10
CommentCure in place versatile low outgassing foam, application life 6 minutes

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application