My Account Menu
My Account Menu

R-2360 RTV SILICONE FOAM

Description

  • Two-part, room temperature curing, silicone foam
  • Platinum cure
  • No odor
  • Yields a low density silicone foam when catalyzed
  • 1:1 Mix Ratio (Part A: Part B)

Applications

  • In shock and vibration dampening situations requiring a light weight, flexible foam with excellent thermal insulation.
  • Applications requiring a low density component for better fuel economy, and other applications where the weight needs to be considered.


Product Details

PROPERTYAVERAGE RESULT
Viscosity40,000 cP (40,000 mPa*s)
Cure SystemPlatinum
Foam Density12 lbs/ft3 (0.19 g/cm3)
Application Life2 minutes
AppearanceWhite
Cure20 minutes / RTV
Mix Ratio1:1
CommentFlexible cure in place low density foam. Application life 2 minutes.

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application