My Account Menu
My Account Menu

CV-1152 CONTROLLED VOLATILITY RTV SILICONE CONFORMAL COATING

Description

  • One-part, flowable, solvent free silicone

Applications

  • For industrial or space applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
  • As a conformal coating on rigid and flexible circuit boards
  • For encapsulating sensitive electronic components such as connectors and switches
  • To provide protection against harsh environments while maintaining good dielectric properties
  • For applications requiring a broader operating temperature range


Product Details

PROPERTYAVERAGE RESULT
Viscosity7,300 cP (7,300 mPa*s)
AppearanceTranslucent
Tack Free Time50 minutes
Cure7 days / RTV
Cure SystemOxime
Mix Ratio1 PART
Refractive Index1.43
Specific Gravity1.01
CommentSoft, very low modulus coating

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn More

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.

Speak with an Expert

Our NuSil team is ready to collaborate with you to find the right silicone solution for your application