My Account Menu
My Account Menu

CV7-2289-1 CONTROLLED VOLATILITY POTTING AND ENCAPSULATING SILICONE ELASTOMER

 

 

  • Two-part, white silicone system
  • 1:1 Mix Ratio (Part A:B)

 

  • For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
  • To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
  • Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
  • For applications requiring a broader operating temperature range


 

PROPERTYAVERAGE RESULT
Cure SystemPlatinum
Viscosity228,750 cP (228,750 mPa*s)
Lap Shear350 psi (2.41 MPa)
Durometer30 Type A
AppearanceWhite
Cure15 minutes / 150 °C
Elongation375 %
Mix Ratio1:1
Tack Free Time40 hours
Tensile700 psi (4.83 MPa)
CommentPrimerless Adhesion