CV-2289-1 CONTROLLED VOLATILITY POTTING AND ENCAPSULATING SILICONE ELASTOMER
Description
- Two-part, white silicone system
- Offers a high tear strength, good physical properties and a broad operating temperature range
- Convenient 1:1 mix ratio (Part A: Part B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For use as an adhesive
- For applications requiring a broader operating temperature range
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Cure System | Platinum |
| Rheology | Flowable / Self Leveling |
| Work Time | 30 minutes |
| Durometer | 34 Type A |
| Appearance | White |
| Cure | 15 minutes / 150 °C |
| Cure Type | Heat Cure / RTV |
| Dielectric Strength | 955 volts/mil (37.25 kV/mm) |
| Elongation | 343 % |
| Lap Shear | 375 psi (2.6 MPa) |
| Mix Ratio | 1:1 |
| Rheology | Flowable / Self Leveling |
| Specific Gravity | 1.12 |
| Tack Free Time | 5 hours |
| Tensile | 720 psi (5.0 MPa) |
| Viscosity | 50,000 cP (50,000 mPa*s) |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
