CV2-2289-1 CONTROLLED VOLATILITY POTTING AND ENCAPSULATING SILICONE ELASTOMER
Description
- Two-part, white silicone system
- Offers medium strength, low modulus, good physical properties and a broad operating temperature range
- 1:1 Mix Ratio (Part A:B)
Applications
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for adhesive applications where a large surface must be covered
- For applications requiring minimal outgassing
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Cure System | Platinum |
| Viscosity | 12,300 cP (12,300 mPa*s) |
| Lap Shear | 300 psi (2.07 MPa) |
| Durometer | 30 Type A |
| Appearance | White |
| Cure | 4 hours / 65 °C |
| Elongation | 250 % |
| Mix Ratio | 1:1 |
| Specific Gravity | 1.1 |
| Tack Free Time | 20 hours |
| Tensile | 450 psi (3.1 MPa) |
| Comment | White, versatile adhesive with short pot life, tack free in 20 hrs Recommended for broad temperature applications. 300 psi (2.1 MPa) primed (CF1-135) lap shear. |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
