CV1-2289-1 CONTROLLED VOLATILITY POTTING AND ENCAPSULATING SILICONE ELASTOMER
Description
- Two-part, white silicone system
- 1:1 Mix Ratio (Part A:B)
Applications
- For applications requiring low outgassing and minimal volatile condensables under extreme operating conditions
- To provide protection of electric components and assemblies against shock, vibration, moisture, dust, chemicals and other environmental hazards
- Ideal for use in potting connectors, cable harness breakouts, molded high-voltage terminals, seals and gaskets due to its high tear strength
- For applications requiring a broader operating temperature range
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Cure System | Platinum |
| Viscosity | 52,500 cP (52,500 mPa*s) |
| Lap Shear | 450 psi (3.1 MPa) |
| Durometer | 30 Type A |
| Appearance | White |
| Cure | 15 minutes / 150 °C |
| Elongation | 350 % |
| Mix Ratio | 1:1 |
| Specific Gravity | 1.1 |
| Tack Free Time | 17 hours |
| Tensile | 750 psi (5.17 MPa) |
| Work Time | 4 hours |
| Comment | White, versatile adhesive with 3 hr pot life. Recommended for broad temperature applications. 450 psi (3.1 MPa), primed (CF1-135) lap shear. |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
