CAT-41 INHIBITOR
Description
- A translucent inhibitor known as Ethynlcyclohexanol
- Amounts used can be varied for optimal processing times
Applications
- For use with platinum catalyzed elastomer systems
Product Details
| PROPERTY | AVERAGE RESULT |
|---|---|
| Appearance | Translucent Yellow |
| Cure | >120 °C |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please Contact NuSil for assistance and recommendations in establishing limits for product specifications.
Speak with an Expert
Our NuSil team is ready to collaborate with you to find the right silicone solution for your application
