My Account Menu
United States
My Account Menu
coatings-dispersions-hero_rdax_1170x548_80.jpg
Silicone coatings and dispersions
Silicone coatings and dispersions for protection in power electronics and biocompatibility in medical devices
Shop Now
NuSil® coatings and dispersions from Avantor® create and protect critical parts or technologies for a wide range of applications in the healthcare, space, aviation, defense, optoelectronics and electronics industries.
NuSil silicone coatings and dispersions are elastomers dispersed in a solvent for ease of application. They can be applied in very thin layers to evenly coat a device or surface or used to create a film or thin-walled part with strong elastomeric properties.
NuSil’s broad range of silicone coatings and dispersions offer numerous options to accommodate processing requirements and functional properties:  
  • One-part or two-part coatings: Our coatings are available as a one-part formulation with no mixing required or two-part configuration offering a longer work time.  
  • Solvent-based or solventless coatings: Solvent-based coatings are ideal for processes where the silicone is applied as a thin film coating. Solvent-less coatings are non-flammable and do not require controlled environments for handling.
  • Special features: NuSil coating options include hydrocarbon-resistance, lubrication, low coefficient of friction and high clarity formulations.
Custom formulations tailored to your needs
nusil-quality-375x235.jpg
NuSil has the expertise, processes, and proprietary technology to customize our silicones based on your unique specifications. We leverage our insight into the unique silicone chemistry and material properties to supply off-the-shelf and custom formulations tailored to your process and end-application performance requirements.
Learn more about how we can help you get the products you need.
Contact us
Product finder
medical-implants-2-375x235.jpg
For long term implanted devices (>29 days) NuSil silicone coatings and dispersions are ideal for dipping and spraying processes that produce thin elastomeric films or forms.
general-healthcare-375x235.jpg
Our portfolio of medical-grade coatings and dispersions offer solutions for devices implanted in the body for < 29 days, such as balloons or external devices like needles.
mars-rover-375x235.jpg
Our space-grade coatings protect against atomic oxygen (AO) while providing resistance to radiation and are used on a variety of spacecraft surfaces and small electronic components.
aerospace-defense-375x235.jpg
NuSil coatings feature fuel and heat-resistant properties that protect critical technology in missiles, rockets, launch equipment and unmanned aerial vehicles.
optoelectronics-375x235.jpg
From extending the operating life of circuit boards to providing stability for elastomers, our conformal coatings protect sensitive elements from moisture, contaminants, shock and temperature extremes.
Related content
Browse additional resources related to silicone conformal coatings.
White papers
Advanced technologies
conformal-coatings-260x190.jpg
Conformal coatings
The NuSil® brand of silicone conformal coatings efficiently and reliably protects electronic components and helps ensure long operating life. Let us put our silicone expertise to work for you.
Learn more
How can we help you?
Whether you’re using NuSil HCRs to extrude tubing or LVEs to encapsulate electronics, NuSil experts will help you determine the right silicone for your application.
Contact us