VWR® Shakers
VWR® shakers are designed for a wide range of application including cell cultures, immunoassays, gels and blots. The microprocessor ensures the speed is slowly increased to avoid splashing and provides accurate and repeatable results. Most models include a timer that can be programmed to a set time and turn off when reaching zero.
Models are available as incubating or open air. Choose orbital, rocking, waving, or reciprocating motions to best meet your application needs. Capacities from microplates up to 150lbs allow you to customize your shaker with over 100 accessory choices.
Product selection
Designed for a wide range of applications including cell cultures that require accurate and repeatable results. The microprocessor control provides consistent, uniform shaking while safely ramping the set speed.
An easy and economical option for all of your rocking needs. Ideal for cell culture and blotting applications. Designed to be used in a variety of environmental conditions.
Innovatively designed to incubate and shake samples. Provides accurate and repeatable results.
Deigned for heating and cooling applications that require consistent and precise high-speed shaking. Innovative on-board help screens make operations simple.