VWR® Hotplates and Stirrers
The VWR® Advanced and Professional Hotplate-Stirrers have been redesigned with safety as a priority. Industry leading heat monitoring and shutoff features promote a safe laboratory environment.
World-Class temperature capabilities coupled with powerful stirring make these models the right choice. All heating models feature a connection for a temperature probe and have calibration functions. Assembled in the USA, including a best-in-class 5 year warranty. Hotplate-Stirrers are available in 7x7", 10x10",and 135mm diameter round top.
Select Professional models when enhanced features such as a timer, temperature limiting function and monitoring the actual versus set temperature are needed and includes a PT1000 RTD temperature probe. Choose the right model for your application.
Product Selection
VWR® Advanced Magnetic Hotplates Stirrers
Advanced magnetic hotplate stirrers are designed to deliver accurate and repeatable results with digitally controlled mixing and superior temperature control. Available in three sizes and a choice of two top plate materials.
VWR® Professional Hotplate-Stirrers
Designed for applications requiring exceptional accuracy, stability and repeatability. These hotplate-stirrers are equipped with superior heating and mixing capabilities. Each include a timer and temperature probe. Available in two sizes.