Performance level 3: High performance, When sensitivity is a must
Protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses
Our Thermo Scientific™ SureSTART™ Performance Level 3 products are packed in clean rooms to protect them against environmental factors and contain the lowest amounts/levels of metals, extractables and leachable; ensuring that you can get the sensitivity and selectivity from your most challenging analytes confidently and consistently.
If you are looking for the ultimate performance from your methods, whilst maintain the highest levels of sample security and integrity, our Performance Level 3 products will meet your needs. Select the products with the three waves.
For analytes that are prone to glass adsorption why not try our GOLD-grade glass technology which has been specifically designed for these more challenging polar analytes, as well as our silanized vials and inserts.
This offering includes Thermo Scientific™ SureStop™ vials, which, when used with 9mm AVCS™ caps, ensure an optimal seal every time. The SureStop™ vial - AVCS™ cap system is specially designed to prevent screw thread vial failures such as cap tilt, cap deformities, dislodged septa or septa push through, thus prevents injection failures and sample evaporation.
Our Performance level 3 septa have the highest purity as they have been conditioned to remove all the low-mid boiling volatiles providing the lowest extractables and leachables within the collection. You can also find within this performance level soft septa options for ultimate needle performance, as these products offer protection for even the most fragile needles with the highest gauge and SPME fibers. You can also find our high temperature compatible septa here.
- All products within this performance level are suitable for GC, HPLC/UHPLC, and nominal mass and high-resolution accurate mass (HR-AM) applications.
- Volume sizes ranging from 0.2 mL to 20 mL covering limited sample volumes to headspace analyses.
- LC/GC Certified, MSCERT and MSCERT+ Kits can be found within performance level 3.
Specification certified
- All Thermo Scientific™ SureSTART™ vials are dimensionally verified.
- Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances.
- Meet all relevant requirements of US, EU, JPN Pharmacopeia.
- Uniform and defect-free
Lot traceable
- 100% traceable through the manufacturing process
- Certificate of Conformance available by lot number upon request
Performance level 3 products include:
SureSTART™ Vials/ Closures
Standard 2 mL Glass: Amber/Clear
- Crimp 2mL /11mm Crimp Caps(Magnetic Steel and Aluminum)
- Screw 2mL (standard and SureSTOP™)/9mm Screw AVCS™ Caps
- Snap 2mL /11mm Snap and Crimp Caps (Magnetic Steel and Aluminum)
GOLD-Grade Glass: Amber/Clear vials & inserts
- 2mL Screw and Crimp GOLD-GRADE (standard and SureSTOP™) / 9mm Screw AVCS™ Caps with 11mm Crimp Caps (Magnetic Steel and Aluminum)
- 2mL Inserts GOLD-Grade
Microvials for < 2 mL samples
- Crimp microvials < 2 mL/ 11mm crimp caps
- Screw microvials < 2 mL/ 9mm screw caps
- Screw total recovery microvials 1.5 mL/ 9mm screw caps
- Screw high recovery microvials 1.7 mL/ 9mm screw caps
- Screw TPX microvials 0.02 mL/ 9mm screw caps
- Snap microvials < 2 mL/ 11mm snap and cirmp caps
- Snap 1.5mL total recovery microvials < 2 mL/ 11mm snap and crimp caps
- Snap high recovery microvials < 2 mL/ 11mm snap and crimp caps
Inserts
Headspace Glass
Vial/Cap. Septa Kits
- HPLC &GC Certified Screw Vial / Cap Kits
2 mL screw /9mm caps (includes SureSTOP™ vials/ AVCS™ caps) - MS Certified Screw Vial/ Cap Kits
2 mL screw /9mm caps (includes SureSTOP™ vials/ AVCS™ caps) - MS Certified+ Screw Pre-cleaned Vial/ Cap Kits
0.2, 1.5, 2mL screw options (includes SureSTOP™ vials/ AVCS™ caps) - TOC Certified Screw and Cap Kits
Certified 40mL screw/ 24mm closures - Certified Screw and Cap Kits
0.2, 2ML glass (includes SureStop™) or TPX and polypropylene or AVCS™ caps with silicone/PTFE septa. Bonded septa options are available.
Crimpers, Decrimpers and Decapping
- Crimpers, Decrimpers and Decapping Pliers
- Manual Crimpers and Decrimpers for Cleanrooms
- Accessories for Programmable Electronic High-Power Crimp Stations