GraphEx Cut Protection
SW® has brought its innovation and hand health mindset to cut-resistant gloves. Our new products are developed in the service of protecting workers and giving them more comfort and confidence to make them more successful every day.
GraphEx® gloves are made with proprietary AxiFybr® yarn—engineered at the molecular level to provide the highest level of cut protection without loss of comfort, fit, or flexibility.
Product Selection
AxiFybr® cut resistant technology in a comfortable 18 gauge liner provides ANSI A4 cut and ANSI 5 abrasion protection. Touchscreen compatible smooth coating.
AxiFybr® cut resistant technology in a comfortable 18 gauge liner provides ANSI A4 cut and ANSI 6 abrasion protection. Touchscreen compatible sandy finish coating.
AxiFybr® technology and proprietary coating provide high cut protection and durability. Rated at ANSI A6 cut and ANSI 6 abrasion. Feel comfort without performance loss.