Cleaning/CE Swabs
In controlled environments, precision is everything.
You can count on Puritan for cleanroom and industrial swabs that are manufactured in an ISO:2008 certified and CE-compliant facility, ensuring the quality of your end product. Whether you work in precision manufacturing, fiber optics, electronics, or biotechnology, you'll always have the right swab for the job. And we're not just talking about foam, cotton, and knitted polyester. You won't find better ESD (electrostatic discharge) safe, anti-static, or chemical-resistant swabs on the market.
This absorbent cotton tipped applicator is produced with high-quality, USP-grade cotton which is carded and coiled onsite to maintain consistency. It is bonded securely to a rigid white birch handle by an aqueous-based adhesive.
This absorbent cotton-tipped applicator is produced with high-quality, USP-grade cotton which is carded and coiled onsite to maintain consistency. It is bonded securely to a sturdy wood handle by an aqueous-based adhesive...
PurSwab® ESD Foam Tipped Swabs
This foam-tipped applicator is produced with reticulated 100ppi foam. It is thermally welded to a static-dissipative solid polypropylene shaft.
This lint-free swab has a large knitted polyester fiber tip that is ultrasonically welded to an injection-molded polypropylene handle. Ideal for cleanroom use and other critical environments.